3-Bromo-5-chloropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43740-0050
43740-0250
TS43740-0050EA
155
USD
TS43740-0050
TS43740-0250
3-Bromo-5-chloropyridine 97%
3-Bromo-5-chloropyridine
Formula:
C₅H₃BrClN Melting Pt: 78.7…80.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114221 CAS Number: 73583-39-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 77.0 to 81.0 °C |
Water | ≤3.00 % |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...