2-Cyanopyrazine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile

43692-0100 43692-0500
TS43692-0100EA 38 USD
TS43692-0100 TS43692-0500
2-Cyanopyrazine 99%
2-Cyanopyrazine
Formula: C₅H₃N₃
MW: 105.10 g/mol
Boiling Pt: 90…93 °C (15 mmHg)
Melting Pt: 20 °C
Density: 1.174 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00049361
CAS Number: 19847-12-2
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5330 to 1.5350 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR