3-Bromo-2-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromo-2-picoline

43686-0010 43686-0050
TS43686-0010EA 89 USD
TS43686-0010 TS43686-0050
3-Bromo-2-methylpyridine 97%
3-Bromo-2-methylpyridine
Formula: C₆H₆BrN
MW: 172.02
Boiling Pt: 65…67 °C (10Torr)
Density: 1.495 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00191224
CAS Number: 38749-79-0

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR