2-Bromo-5-iodopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43685-0050
43685-0250
TS43685-0050EA
74
USD
TS43685-0050
TS43685-0250
2-Bromo-5-iodopyridine 97%
2-Bromo-5-iodopyridine
Formula:
C₅H₃BrIN Melting Pt: 123…123.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095201 CAS Number: 73290-22-9 |
Specification Test Results
Appearance | White to yellow to light pink to light brown Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 121 to 125 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...