3-Amino-1-Boc-piperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate

43660-0010 43660-0050
TS43660-0010EA 247 USD
TS43660-0010 TS43660-0050
3-Amino-1-Boc-piperidine 97%
3-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861219
CAS Number: 184637-48-7

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR