Nortropinone hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
8-Azabicyclo[3.2.1]octan-3-one hydrochloride
43612-0010
43612-0050
TS43612-0010EA
109
USD
TS43612-0010
TS43612-0050
Nortropinone hydrochloride 97%
Nortropinone hydrochloride
Formula:
C₇H₁₂ClNO MW: 161.63 g/mol Melting Pt: 194.7…195.8 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03613582 CAS Number: 25602-68-0 |
Specification Test Results
Appearance | White to off-white to light brown Powder or solid |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 to ≤104.0 % |
GC | ≥98.0 % |
Specific optical rotation | +12° to +15° (20°C, 589 nm) (Neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...