2-Fluoro-4-pyridineboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoropyridine-4-boronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

43593-0010 43593-0050
TS43593-0010EA 165 USD
TS43593-0010 TS43593-0050
2-Fluoro-4-pyridineboronic acid pinacol ester 95%
2-Fluoro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
Melting Pt: 47.9…51.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06798253
CAS Number: 458532-86-0

Order Now

Specification Test Results

Appearance Yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 47 to 54 °C
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR