(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulfonate salt 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43583-0010
43583-0050
TS43583-0010EA
113
USD
TS43583-0010
TS43583-0050
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulfonate salt 97%
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulfonate salt
Formula:
C₁₁H₁₇NO₄S Melting Pt: 137.9…142.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066238 CAS Number: 104530-79-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white to light brown Crystals or solid |
Infrared spectrum | Conforms |
Elemental Analysis | 5.2 to 5.6 % N |
Specific optical rotation | -3° to -5° (20°C, 589 nm) (c=1, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...