1-Methyl-1H-indazol-3-amine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Amino-1-methylindazole, 1-methyl-1H-indazole-3-ylamine, 3-Amino-1-methyl-1H-indazole

43572-0010 43572-0050
TS43572-0010EA 69 USD
TS43572-0010 TS43572-0050
1-Methyl-1H-indazol-3-amine 95%
1-Methyl-1H-indazol-3-amine
Formula: C₈H₉N₃
MDL Number: MFCD09054755
CAS Number: 60301-20-4

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 100 °C
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR