5-Iodo-1H-indazole 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43571-0010
43571-0050
TS43571-0010EA
200
USD
TS43571-0010
TS43571-0050
5-Iodo-1H-indazole 95%
5-Iodo-1H-indazole
Formula:
C₇H₅IN₂ MW: 244.0300 g/mol Melting Pt: 161…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD07781642 CAS Number: 55919-82-9 |
Specification Test Results
Appearance | Yellow to brown to purple Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 151 to 156 °C |
HPLC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...