tert-Butyl-4-oxoazepane-1-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tert-butyl 4-oxoazepane-1-carboxylate, 4-Oxoazepane-1-carboxylic acid tert-butyl ester, N-BOC-Hexahydro-1H-azepin-4-one, tert-Butyl 4-oxoazepane-1-carboxylate

43554-0010 43554-0050
TS43554-0010EA 133 USD
TS43554-0010 TS43554-0050
tert-Butyl-4-oxoazepane-1-carboxylate 97%
tert-Butyl-4-oxoazepane-1-carboxylate
Formula: C₁₁H₁₉NO₃
MW: 213.277 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03788435
CAS Number: 188975-88-4

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR