6-Bromonicotinic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromopyridine-3-carboxylic acid, 2-Bromo-5-pyridinecarboxylic acid, 6-Bromo-3-pyridinecarboxylic acid

43540-0010 43540-0100
TS43540-0010EA 41 USD
TS43540-0010 TS43540-0100
6-Bromonicotinic acid 96%
6-Bromonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 179…181 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01927100
CAS Number: 6311-35-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR