1,3-Dicyclohexylbenzimidazolium chloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Dicyclohexyl-1H-benzo[d]imidazol-3-ium chloride, 1,3-Dicyclohexyl-1H-benzimidazol-3-ium chloride

43509-0010 43509-5000
TS43509-0010EA 163 USD
TS43509-0010 TS43509-5000
1,3-Dicyclohexylbenzimidazolium chloride 95%
1,3-Dicyclohexylbenzimidazolium chloride
Formula: C₁₉H₂₇ClN₂
MW: 318.89 g/mol
Melting Pt: 209.9…211.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08705248
CAS Number: 1034449-15-4

Order Now

Specification Test Results

Appearance White to grey to tan Powder
Infrared spectrum Conforms
Elemental Analysis 8.25 to 9.31 % (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR