1-Boc-4-chloropiperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

43501-0010 43501-0050
TS43501-0010EA 150 USD
TS43501-0010 TS43501-0050
1-Boc-4-chloropiperidine 97%
1-Boc-4-chloropiperidine
Formula: C₁₀H₁₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD04115040
CAS Number: 154874-94-9

Order Now

Specification Test Results

Appearance Colorless to white Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR