6-Hydroxypicolinic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Hydroxy-2-pyridinecarboxylic acid, 6-Hydroxypyridine-2-carboxylic acid

43482-0050 43482-0250
TS43482-0050EA 62 USD
TS43482-0050 TS43482-0250
6-Hydroxypicolinic acid 95%
6-Hydroxypicolinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 273…276 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00192220
CAS Number: 19621-92-2

Order Now

Specification Test Results

Appearance White to yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR