4-Bromo-2-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

43480-0050 43480-0250
TS43480-0050EA 99 USD
TS43480-0050 TS43480-0250
4-Bromo-2-chloropyridine 97%
4-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
Density: 1.734 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD03840756
CAS Number: 73583-37-6

Order Now

Specification Test Results

Appearance Clear colorless to white to yellow to orange Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR