1-Methylbenzimidazole 99%

Supplier: Thermo Scientific Chemicals
Warning

43442-0050 43442-0250
TS43442-0050EA 67 USD
TS43442-0050 TS43442-0250
1-Methylbenzimidazole 99%
1-Methylbenzimidazole
Formula: C₈H₈N₂
Boiling Pt: 154 °C (16hPa)
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00192275
CAS Number: 1632-83-3

Order Now

Specification Test Results

Appearance White to pink solid
Infrared spectrum Authentic
Melting point 55 to 60 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR