Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: methyl 4-oxotetrahydro-2H-thiopyran-3-carboxylate, methyl-4-oxotetrahydro-2H-thiopyran-3-carboxylate, Methyl 4-oxothiane-3-carboxylate

43429-0010 43429-0050
TS43429-0010EA 190 USD
TS43429-0010 TS43429-0050
Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate 95%
Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate
Formula: C₇H₁₀O₃S
MW: 174.214 g/mol
Boiling Pt: 134 °C (10Torr)
Storage Temperature: Ambient
MDL Number: MFCD00203485
CAS Number: 4160-61-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or solid
Infrared spectrum Conforms
GC ≥94.0 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR