2-Bromo-5-methylthiophene 95% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol

Stabilization: Stabilised with copper and sodium hydrogen carbonate (0.1:0.4)%

43420-0010 43420-0050
TS43420-0010EA 41 USD
TS43420-0010 TS43420-0050
2-Bromo-5-methylthiophene 95% stabilized
2-Bromo-5-methylthiophene
Formula: C₅H₅BrS
MW: 177.06 g/mol
Boiling Pt: 65 °C (15 mmHg)
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130103
CAS Number: 765-58-2

Order Now

Specification Test Results

Appearance Yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
GC ≥94.0 %
Stabilizer 0.1 % Cu and 0.4 % NaHCO3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR