3-Cyanophenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 3-Cyanobenzeneboronic acid pinacol ester

43379-0010 43379-0050
TS43379-0010EA 89 USD
TS43379-0010 TS43379-0050
3-Cyanophenylboronic acid pinacol ester 97%
3-Cyanophenylboronic acid pinacol ester
Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
Melting Pt: 80…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03789262
CAS Number: 214360-46-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 78 to 82 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR