3-Aminomethyltetrahydrofuran 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Tetrahydrofuran-3-yl)methanamine, 3-(Aminomethyl)oxolane, (tetrahydrofuran-3-yl)methanamine, Oxolan-3-ylmethanamine, 3-(Aminomethyl)tetrahydrofuran

43300-0010 43300-0050
TS43300-0010EA 310 USD
TS43300-0010 TS43300-0050
3-Aminomethyltetrahydrofuran 97%
3-Aminomethyltetrahydrofuran
Formula: C₅H₁₁NO
MW: 101.149 g/mol
Boiling Pt: 65 °C (15Torr)
Storage Temperature: Ambient
MDL Number: MFCD08234925
CAS Number: 165253-31-6
UN: 2924
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Residue after ignition ≤0.1 %
pH 9.5 to 11.0 (5% at 25°C)
Titration with HClO4 ≥99 %
Iron (Fe) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR