5-Amino-2-methylbenzothiazole 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylbenzo[d]thiazol-5-amine, 2-Methyl-1,3-benzothiazol-5-amine

43276-0010 43276-0050
TS43276-0010EA 42 USD
TS43276-0010 TS43276-0050
5-Amino-2-methylbenzothiazole 95%
5-Amino-2-methylbenzothiazole
Formula: C₈H₈N₂S
MW: 164.23 g/mol
Melting Pt: 101…103.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00226291
CAS Number: 13382-43-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 105 °C
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR