5-Bromo-2-morpholin-1-ylpyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-(4-morpholino)pyrimidine, 4-(5-Bromopyrimidin-2-yl)morpholine

43270-0050 43270-0250
TS43270-0050EA 125 USD
TS43270-0050 TS43270-0250
5-Bromo-2-morpholin-1-ylpyrimidine 97%
5-Bromo-2-morpholin-1-ylpyrimidine
Formula: C₈H₁₀BrN₃O
MW: 244.1 g/mol
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00483251
CAS Number: 84539-22-0

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 86 to 93 °C
GC ≥96.0 %
Specific optical rotation -63° to -67° (20 °C, 589 nm) (c=4, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR