5-Fluoro-2-hydroxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

43258-0050 43258-0250
TS43258-0050EA 177 USD
TS43258-0050 TS43258-0250
5-Fluoro-2-hydroxypyridine 97%
5-Fluoro-2-hydroxypyridine
Formula: C₅H₄FNO
MW: 113.09 g/mol
Melting Pt: 152…153 °C
Storage Temperature: Ambient
MDL Number: MFCD03092918
CAS Number: 51173-05-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 155 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR