2-Amino-4-phenylthiazole 97%

Supplier: Thermo Scientific Chemicals
Danger

43249-0010 43249-0050
TS43249-0010EA 50 USD
TS43249-0010 TS43249-0050
2-Amino-4-phenylthiazole 97%
2-Amino-4-phenylthiazole
Formula: C₉H₈N₂S
Melting Pt: 150…152.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00039680
CAS Number: 2010-06-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 149 to 153 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR