2-Methyl-4-(4-morpholinyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-4-(4-morpholinyl)benzenamine

43246-0010 43246-0050
TS43246-0010EA 170 USD
TS43246-0010 TS43246-0050
2-Methyl-4-(4-morpholinyl)aniline 97%
2-Methyl-4-(4-morpholinyl)aniline
Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD10686817
CAS Number: 581-00-0

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 89 to 93 °C
HPLC ≥96.0 %
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR