6-Iodo-1H-indazole 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Iodoindazole

43242-0010 43242-0050
TS43242-0010EA 281 USD
TS43242-0010 TS43242-0050
6-Iodo-1H-indazole 97%
6-Iodo-1H-indazole
Formula: C₇H₅IN₂
MW: 244.0300 g/mol
Melting Pt: 208…211 °C
Storage Temperature: Ambient
MDL Number: MFCD04114695
CAS Number: 261953-36-0

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 206 to 211 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR