1-Boc-3-oxopiperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-butyl 3-oxopiperazine-1-carboxylate, tert-butyl 3-oxo-1-piperazinecarboxylate, 1-Boc-3-Oxopiperazine, 4-N-BOC-2-oxo-piperazine, 4-Boc-2-Oxopiperazine

43233-0010 43233-0050
TS43233-0010EA 70 USD
TS43233-0010 TS43233-0050
1-Boc-3-oxopiperazine 97%
1-Boc-3-oxopiperazine
Formula: C₉H₁₆N₂O₃
MW: 200.238 g/mol
Melting Pt: 160…163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02181069
CAS Number: 76003-29-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 164 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR