3-Quinolineboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Quinoline-3-boronic acid pinacolate, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline, Quinoline-3-boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-quinolyl)-1,3,2-dioxaborolane
43211-0010
43211-0050
TS43211-0010EA
76
USD
TS43211-0010
TS43211-0050
3-Quinolineboronic acid pinacol ester 97%
3-Quinolineboronic acid pinacol ester
Formula:
C₁₅H₁₈BNO₂ Melting Pt: 55…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD05155218 CAS Number: 171364-85-5 |
Specification Test Results
Appearance | White to light brown Powder or solid |
Infrared spectrum | Conforms |
Melting point | 55 to 60 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...