4-Bromo-2-pyridone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromopyridin-2-ol, 4-bromo-1,2-dihydropyridin-2-one, 4-Bromo-2-hydroxypyridine

43209-0010 43209-0050
TS43209-0010EA 180 USD
TS43209-0010 TS43209-0050
4-Bromo-2-pyridone 97%
4-Bromo-2-pyridone
Formula: C₅H₄BrNO
MW: 174.00 g/mol
Melting Pt: 203…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00234041
CAS Number: 36953-37-4

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR