(R)-(+)-1-Boc-3-methylpiperazine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3R)-3-Methylpiperazine, N1-BOC protected, (R)-1-Boc-3-Methylpiperazine, (R)-1-Boc-3-methyl-piperazine

43205-0010 43205-0050
TS43205-0010EA 69 USD
TS43205-0010 TS43205-0050
(R)-(+)-1-Boc-3-methylpiperazine 99%
(R)-(+)-1-Boc-3-methylpiperazine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 44.4…46.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02683205
CAS Number: 163765-44-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 43 to 47 °C
GC ≥98.5 %
Specific optical rotation +14° to +18° (20 °C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR