2-Bromo-5-formylthiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromothiazole-5-carboxaldehyde

43101-0010 43101-0050
TS43101-0010EA 151 USD
TS43101-0010 TS43101-0050
2-Bromo-5-formylthiazole 97%
2-Bromo-5-formylthiazole
Formula: C₄H₂BrNOS
MW: 192.03 g/mol
Melting Pt: 112…113.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788567
CAS Number: 464192-28-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 114 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR