1-Boc-4-chlorosulfonylpiperidine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Boc-4-Chlorosulphonylpiperidine, 1-BOC-4-PIPERIDINEsulphONYL CHLORIDE, tert-Butyl 4-(chlorosulphonyl)piperidine-1-carboxylate, 4-CHLOROsulphONYL-PIPERIDINE-1-CARBOXYLIC acid TERT-BUTYL ESTER

43099-0010 43099-0050
TS43099-0010EA 264 USD
TS43099-0010 TS43099-0050
1-Boc-4-chlorosulfonylpiperidine 97%
1-Boc-4-chlorosulfonylpiperidine
Formula: C₁₀H₁₈ClNO₄S
MW: 283.767 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861750
CAS Number: 782501-25-1
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to pale yellow Powder
Infrared spectrum Conforms
Melting point 82 to 87 °C
Elemental Analysis 4.76 to 5.11 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR