Propylene oxide 99.5%, AcroSeal™, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide

43086-1000 43086-8000
TS43086-1000EA 27 USD
TS43086-1000 TS43086-8000
Propylene oxide 99.5%, AcroSeal™, Extra Pure
Propylene oxide
Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 34 °C
Melting Pt: –112 °C
Density: 0.83 g/cm³
Flash Pt: –37 °C
Storage Temperature: Ambient
MDL Number: MFCD00005126
CAS Number: 75-56-9
UN: 1280
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=443494ff0620e18a4c5de7f3f4cd87d6e5eec44c&vid=33bc95a664261fbc904256d1b87a1972dde5dcd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3650 to 1.3670 (20°C, 589 nm)
Water ≤0.02 %
GC ≥99.4 %
Color scale ≤10 APHA
Aldehyde (C3H6O) ≤0.01

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR