2-Chloro-3-pyridineboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-3-boronic acid pinacol ester

43066-0010 43066-0050
TS43066-0010EA 30 USD
TS43066-0010 TS43066-0050
2-Chloro-3-pyridineboronic acid pinacol ester 97%
2-Chloro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
Melting Pt: 34…39 °C
Storage Temperature: Ambient
MDL Number: MFCD04039873
CAS Number: 452972-11-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR