4-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

43061-0010 43061-0050
TS43061-0010EA 56 USD
TS43061-0010 TS43061-0050
4-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MW: 262.11 g/mol
Melting Pt: 82…83.2 °C
Storage Temperature: Ambient
MDL Number: MFCD02179438
CAS Number: 171364-80-0

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR