3-Amino-4-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

42986-0050 42986-0250
TS42986-0050EA 59 USD
TS42986-0050 TS42986-0250
3-Amino-4-methylpyridine 97%
3-Amino-4-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00128871
CAS Number: 3430-27-1

Order Now

Specification Test Results

Appearance Brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 100 to 108 °C
HPLC ≥96.0 %
Uniformity coefficient ≤2.00
Moisture holding capacity 60 to 70 % (H)
Total volume capacity ≥2.60 eq/l (H)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR