2-Pyrimidinecarbonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Cyanopyrimidine

42982-0050 42982-0250
TS42982-0050EA 37 USD
TS42982-0050 TS42982-0250
2-Pyrimidinecarbonitrile 98%
2-Pyrimidinecarbonitrile
Formula: C₅H₃N₃
MW: 105.1 g/mol
Melting Pt: 37…41 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00160513
CAS Number: 14080-23-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or low melting solid
Infrared spectrum Conforms
Melting point 35 to 42 °C
GC ≥97.5 %
Specific optical rotation +96° to +107° (20 °C, 589 nm) (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR