3,6-Dihydrothiopyran-4-boronic acid pinacol ester

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran, 2-(3,6-Dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

42961-0010 42961-0050
TS42961-0010EA 196 USD
TS42961-0010 TS42961-0050
3,6-Dihydrothiopyran-4-boronic acid pinacol ester
3,6-Dihydrothiopyran-4-boronic acid pinacol ester
Formula: C₁₁H₁₉BO₂S
MW: 226.141 g/mol
Melting Pt: 42…44.5 °C
Storage Temperature: Ambient
MDL Number: MFCD10700154
CAS Number: 862129-81-5

Order Now

Specification Test Results

Appearance Orange to brown Low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR