1H-Benzimidazole-4-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42951-0010
42951-0050
TS42951-0010EA
321
USD
TS42951-0010
TS42951-0050
1H-Benzimidazole-4-carboxylic acid 97%
1H-Benzimidazole-4-carboxylic acid
Formula:
C₈H₆N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01823426 CAS Number: 46006-36-4 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to light brown Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...