2-Amino-6-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

42950-0010 42950-0050
TS42950-0010EA 28 USD
TS42950-0010 TS42950-0050
2-Amino-6-chloropyridine 97%
2-Amino-6-chloropyridine
Formula: C₅H₅ClN₂
Melting Pt: 72…73.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00234068
CAS Number: 45644-21-1

Order Now

Specification Test Results

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 69 to 75 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR