2-Chloropyrimidine-5-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

42947-0010 42947-0050
TS42947-0010EA 288 USD
TS42947-0010 TS42947-0050
2-Chloropyrimidine-5-carboxylic acid 97%
2-Chloropyrimidine-5-carboxylic acid
Formula: C₅H₃ClN₂O₂
MW: 158.5400 g/mol
Melting Pt: 125…127.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06739036
CAS Number: 374068-01-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR