4-Methyl-2-thiazolecarboxaldehyde 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-THIAZOLECARBOXALDEHYDE, 4-METHYL-, 4-Methylthiazole-2-carbaldehyde, 4-Methyl-1,3-thiazole-2-carboxaldehyde

42935-0010 42935-0050
TS42935-0010EA 59 USD
TS42935-0010 TS42935-0050
4-Methyl-2-thiazolecarboxaldehyde 97%
4-Methyl-2-thiazolecarboxaldehyde
Formula: C₅H₅NOS
MW: 127.161 g/mol
Boiling Pt: 90…92 °C (12mbar)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00022450
CAS Number: 13750-68-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
GC ≥96.0 %
pH 5.5 to 7.5 (10 % solution)
Titration with HClO4 ≥98.5 % (On dry substance)
Sulfate (SO4) ≤0.1 %
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR