1-Methyl-1H-indazole-4-carboxaldehyde 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-1H-indazole-4-carbaldehyde

42934-0010 42934-0050
TS42934-0010EA 278 USD
TS42934-0010 TS42934-0050
1-Methyl-1H-indazole-4-carboxaldehyde 95%
1-Methyl-1H-indazole-4-carboxaldehyde
Formula: C₉H₈N₂O
MW: 160.18 g/mol
Melting Pt: 77.5…79 °C
Storage Temperature: Ambient
MDL Number: MFCD10699167
CAS Number: 1053655-56-3

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 76 to 80 °C
HPLC ≥94.0 %
Sulfate (SO4) ≤0.05 %
Ammonium (NH4) ≤0.01 %
Arsenic (As) ≤1 ppm
Copper (Cu) ≤0.002 %
Iron (Fe) ≤0.005 %
Lead (Pb) ≤0.002 %
Nickel (Ni) ≤0.002 %
Insoluble matter ≤0.025 % (in water and ppt by NH4OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR