DL-Pantolactone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, DL-Pantoyl Lactone, 3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one, DL-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone

42759-0250 42759-1000
TS42759-0250EA 57 USD
TS42759-0250 TS42759-1000
DL-Pantolactone 97%
DL-Pantolactone
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 246 °C (760 mmHg)
Melting Pt: 74…78 °C
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00064333
CAS Number: 79-50-5

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤0.5 %
GC ≥96.0 %
UV at 240 nm A: ≤0.05
Titration after Ion exchange ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR