3-(2-Bromoethyl)-1H-indole 95%

Supplier: Thermo Scientific Chemicals
Danger

42691-0010 42691-0050
TS42691-0010EA 48 USD
TS42691-0010 TS42691-0050
3-(2-Bromoethyl)-1H-indole 95%
3-(2-Bromoethyl)-1H-indole
Formula: C₁₀H₁₀BrN
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00130155
CAS Number: 3389-21-7
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 95 to 101 °C
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR