2-Indancarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dihydro-1H-indene-2-carboxylic acid, Indan-2-carboxylic acid

42636-0010 42636-0050
TS42636-0010EA 155 USD
TS42636-0010 TS42636-0050
2-Indancarboxylic acid 98%
2-Indancarboxylic acid
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00085095
CAS Number: 25177-85-9

Order Now

Specification Test Results

Appearance White to cream to brown Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 132 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure
Sulfated ash ≤0.1 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR