(6-Bromo-2-pyridinyl)methanol 96%

Supplier: Thermo Scientific Chemicals
Warning

42631-0010 42631-0050
TS42631-0010EA 68 USD
TS42631-0010 TS42631-0050
(6-Bromo-2-pyridinyl)methanol 96%
(6-Bromo-2-pyridinyl)methanol
Formula: C₆H₆BrNO
Melting Pt: 34…39 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04039884
CAS Number: 33674-96-3

Order Now

Specification Test Results

Appearance White to yellow Solid
Infrared spectrum Conforms
Melting point 34 to 41 °C
GC ≥95 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR