Benzyl-4-oxo-1-piperidinecarboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Cbz-Piperidin-4-one, 1-Carbobenzoxy-4-piperidone, 1-Cbz-4-piperidone, 4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester, 1-Benzyloxycarbonyl-4-piperidine
42596-0050
42596-0250
TS42596-0050EA
34
USD
TS42596-0050
TS42596-0250
Benzyl-4-oxo-1-piperidinecarboxylate 97%
Benzyl-4-oxo-1-piperidinecarboxylate
Formula:
C₁₃H₁₅NO₃ MW: 233.27 g/mol Boiling Pt: 114…140 °C (.3 mmHg) Density: 1.185 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00673144 CAS Number: 19099-93-5 |
Specification Test Results
Appearance | Colorless to white to light yellow to light orange Powder or solid or liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...