2-Cyano-6-methylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42592-0010
42592-0050
TS42592-0010EA
40
USD
TS42592-0010
TS42592-0050
2-Cyano-6-methylpyridine 97%
2-Cyano-6-methylpyridine
Formula:
C₇H₆N₂ Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190586 CAS Number: 1620-75-3 |
Specification Test Results
Appearance | White to orange to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 68 to 73 °C |
GC | ≥96 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...